Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methyl 10-undecenoate

Catalog No. 115126500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-512-6500ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-512-6500ML Supplier Sigma Aldrich Supplier No. 115126500ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methyl 10-undecenoate

Specifications

CAS 111-81-9
Density 0.882 g/mL (at 25°C (literature))
Boiling Point 245°C (lit.)
Molecular Formula C12H22O2
Refractive Index n20/D 1.437 (literature)
Linear Formula CH2=CH(CH2)8 CO2CH3
MDL Number MFCD00016689
Quantity 1 g
Synonym 10-Undecenoic acid methyl ester; Undecylenic acid methyl ester
Molecular Weight (g/mol) 198.3
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.