accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 111-95-5

IUPAC Name: bis(2-methoxyethyl)amine
Synonyms: bis(2-methoxyethyl)amine
Molecular Weight (g/mol): 133.19
Molecular Formula: C6H15NO2
InChi Key: IBZKBSXREAQDTO-UHFFFAOYSA-N
SMILES: COCCNCCOC
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)