Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bis(2-methoxyethyl)amine 98.0+%, TCI America™
SDP

Catalog No. B138325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B138325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B138325ML Supplier TCI America Supplier No. B138325ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 111-95-5
Molecular Formula C6H15NO2
Molecular Weight (g/mol) 133.191
MDL Number MFCD00025906
InChI Key IBZKBSXREAQDTO-UHFFFAOYSA-N
Synonym 2,2′C-Dimethoxydiethylamine
PubChem CID 2383
IUPAC Name 2-methoxy-N-(2-methoxyethyl)ethanamine
SMILES COCCNCCOC

Specifications

Color Yellow
Boiling Point 172°C
Quantity 25 mL
UN Number 2734
Formula Weight 133.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Bis(2-methoxyethyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.