accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaHg

CAS RN 11110-52-4

IUPAC Name: mercury sodium
Synonyms: mercury sodium
Molecular Weight (g/mol): 223.58
Molecular Formula: HgNa
InChi Key: MJGFBOZCAJSGQW-UHFFFAOYSA-N
SMILES: [Na].[Hg]
Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results