Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium mercury amalgam, ca. 20% sodium

Catalog No. AC388460500
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC388460500 50 g Glass bottle
AC388460100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC388460500 Supplier Thermo Scientific Chemicals Supplier No. 388460500
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 11110-52-4 | HgNa | 223.582 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11110-52-4
Molecular Formula HgNa
Molecular Weight (g/mol) 223.582
MDL Number MFCD00210362
InChI Key MJGFBOZCAJSGQW-UHFFFAOYSA-N
Synonym sodium amalgam, sodium mercury amalgam, sodium mercury, sodium-amalgam, mercury sodium, sodium mercury amalgam, beads, na 10 %, sodium mercury amalgam, beads trace metals basis, na 5 %, sodium mercury amalgam, beads trace metals basis, na 20 %
PubChem CID 9837419
IUPAC Name mercury;sodium
SMILES [Na].[Hg]

Specifications

Color Gray
Quantity 50 g
Packaging Glass bottle
Form Pieces
Merck Index 15, 8713
Chemical Name or Material Sodium mercury amalgam
Hazard Category Danger
Hazard Statement GHS H Statement
In contact with water releases flammable gases which may ignite spontaneously. Fatal if inhaled. Very toxic to aquatic life with long lasting effects. Causes severe skin burns and eye damage. Causes damage to organs through prolonged or repeated exposure. May damage fertility or the unborn child. Reacts violently with water.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Immediately call a POISON CENTER or doctor/physician.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.