accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1112-37-4

IUPAC Name: diethylphosphinoyl chloride
Synonyms: diethylphosphinoyl chloride
Molecular Weight (g/mol): 140.55
Molecular Formula: C4H10ClOP
InChi Key: KGENPKAWPRUNIG-UHFFFAOYSA-N
SMILES: CCP(Cl)(=O)CC
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results