accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1112459-82-1

IUPAC Name: 1-{4-[(thiophen-2-yl)methyl]phenyl}methanamine hydrochloride
Synonyms: 1-[4-(thiophen-2-ylmethyl)phenyl]methanamine hydrochloride
Molecular Weight (g/mol): 239.76
Molecular Formula: C12H14ClNS
InChi Key: BRDMSBXGZSSVJT-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC=C(CC2=CC=CS2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results