accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CSHNH2OHO

CAS RN 1113-41-3

IUPAC Name: 2-amino-3-methyl-3-sulfanylbutanoic acid
Synonyms: (+-)-penicillamine penicillamine DL- DMC DL-3-mercaptovaline pencillamine β-mercaptovaline 3,3-dimethylcysteine D,L-penicillamine DL-β-mercaptovaline
Molecular Weight (g/mol): 149.21
Molecular Formula: C5H11NO2S
InChi Key: VVNCNSJFMMFHPL-UHFFFAOYNA-N
SMILES: CC(C)(S)C(N)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
  • (2)
Quantity 
  • (3)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)