Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Penicillamine 98.0+%, TCI America™
SDP

Catalog No. P13701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P13701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P13701G Supplier TCI America Supplier No. P13701G

Specifications

CAS 1113-41-3
Color White
Molecular Formula C5H11NO2S
Quantity 1 g
MDL Number MFCD00064303
Synonym l-penicillamine, 3-mercapto-l-valine, penicillamine l-form, l-+-beta-mercaptovaline, l-pen, 2r-2-amino-3-methyl-3-sulfanylbutanoic acid, 3-sulfanyl-l-valine, h-pen-oh, unii-no1yib7oig, 3,3-dimethyl-l-cysteine
InChI Key VVNCNSJFMMFHPL-GSVOUGTGSA-N
SMILES CC(C)(C(C(=O)O)N)S
IUPAC Name (2R)-2-amino-3-methyl-3-sulfanylbutanoic acid
Molecular Weight (g/mol) 149.208
PubChem CID 92863
ChEBI CHEBI:50869
Formula Weight 149.21
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Penicillamine
Show More Show Less
RTECSNumber YV9445500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.