accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CBH3CH3CCH3CH3

CAS RN 1113-78-6

IUPAC Name: tris(butan-2-yl)borane
Synonyms: tris(sec-butyl)borane
Molecular Weight (g/mol): 182.16
Molecular Formula: C12H27B
InChi Key: YUPAWYWJNZDARM-UHFFFAOYNA-N
SMILES: CCC(C)B(C(C)CC)C(C)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results