Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tri-sec-butylborane, 1M Solution in Tetrahydrofuran, Thermo Scientific™

Catalog No. AC176708000
Change view
Click to view available options
Quantity:
800 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC176708000 800 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC176708000 Supplier Thermo Scientific Chemicals Supplier No. 176708000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1113-78-6
Molecular Formula C12H27B
Molecular Weight (g/mol) 182.158
InChI Key YUPAWYWJNZDARM-UHFFFAOYSA-N
Synonym tri-sec-butylborane, tri-sec-butylborane solution, tris sec-butyl borane, borane, tris 1-methylpropyl, tri-s-butylboron, trisec-butylborane, tri-s-butyl-borane, tri-sec butylborane, tri butan-2-yl borane, tris 1-methylpropyl borane
PubChem CID 102559
IUPAC Name tri(butan-2-yl)borane
SMILES B(C(C)CC)(C(C)CC)C(C)CC

Specifications

Density 0.79
Linear Formula [C2H5CH(CH3)]3B
Quantity 800 mL
Formula Weight 182.16
Chemical Name or Material Tri-sec-butylborane
Hazard Category AIR SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.