accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCCH3CH3H3CCH3H3CCCH3CH3H3CCH3ClClTi

CAS RN 11136-36-0

IUPAC Name: titanium(4+) bis(pentamethylcyclopenta-1,3-dien-1-ide) dichloride
Synonyms: titanium(4+) bis(pentamethylcyclopenta-1,3-dien-1-ide) dichloride
Molecular Weight (g/mol): 389.23
Molecular Formula: C20H30Cl2Ti
InChi Key: ZBFBXTFQCKIUHU-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Ti+4].CC1=[C-]C(C)(C)C(C)=C1C.CC1=[C-]C(C)(C)C(C)=C1C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results