accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNH

CAS RN 111373-03-6

IUPAC Name: 4-(piperazin-1-yl)benzonitrile
Synonyms: 4-(piperazin-1-yl)benzonitrile
Molecular Weight (g/mol): 187.25
Molecular Formula: C11H13N3
InChi Key: DJJNYEXRPRQXPD-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(C=C1)N1CCNCC1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)