Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(2-Cyanophenyl)piperazine, 97%, Thermo Scientific™

Catalog No. AAL1939214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1939214 25 g
AAL1939206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1939214 Supplier Thermo Scientific Chemicals Supplier No. L1939214

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111373-03-6
Molecular Formula C11H13N3
Molecular Weight (g/mol) 187.246
MDL Number MFCD00040793
InChI Key FRICBZWJFIRJOB-UHFFFAOYSA-N
Synonym 1-2-cyanophenyl piperazine, 2-piperazin-1-yl benzonitrile, benzonitrile, 2-1-piperazinyl, 2-1-piperazino benzonitrile, 2-piperazin-1-yl-benzonitrile, 1-2-cyanophenyl-piperazine, 2-piperazinylbenzenecarbonitrile, pubchem9347, 2-piperazinobenzonitrile, d02yfz
PubChem CID 2735875
IUPAC Name 2-piperazin-1-ylbenzonitrile
SMILES C1CN(CCN1)C2=CC=CC=C2C#N

Specifications

Density 1.115
Boiling Point 314°C to 315°C
Flash Point >100°C (212°F)
Refractive Index 1.59
Quantity 25 g
UN Number UN2922
Beilstein 785973
Sensitivity Moisture sensitive
Formula Weight 187.25
Percent Purity 97%
Chemical Name or Material 1-(2-Cyanophenyl)piperazine
Show More Show Less
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.