accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1114-51-8

IUPAC Name: N,N-diethylpropanamide
Synonyms: propanamide, N,N-diethyl-
Molecular Weight (g/mol): 129.2
Molecular Formula: C7H15NO
InChi Key: YKOQQFDCCBKROY-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results