Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Diethylpropionamide 98.0+%, TCI America™
SDP

Catalog No. D052325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D052325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D052325ML Supplier TCI America Supplier No. D052325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1114-51-8
Molecular Formula C7H15NO
Molecular Weight (g/mol) 129.20
MDL Number MFCD00009302
InChI Key YKOQQFDCCBKROY-UHFFFAOYSA-N
PubChem CID 66191
IUPAC Name N,N-diethylpropanamide
SMILES CCN(CC)C(=O)CC

Specifications

Color Yellow
Boiling Point 191°C
Quantity 25 mL
Formula Weight 129.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N,N-Diethylpropionamide
RTECSNumber UE3567500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.