accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COON

CAS RN 1116-98-9

IUPAC Name: tert-butyl 2-cyanoacetate
Synonyms: tert-butyl 2-cyanoacetate
Molecular Weight (g/mol): 141.17
Molecular Formula: C7H11NO2
InChi Key: BFNYNEMRWHFIMR-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CC#N
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)