Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich tert-Butyl cyanoacetate

Catalog No. 2844010MLF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
28-440-10MLF 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 28-440-10MLF Supplier Sigma Aldrich Supplier No. 2844010MLF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

tert-Butyl cyanoacetate

Specifications

CAS 1116-98-9
Density 0.988 g/mL (at 20°C (literature))
Boiling Point 40°C to 42°C (0.1 mmHg)
Molecular Formula C7H11NO2
Refractive Index n20/D 1.420
Linear Formula NCCH2COOC(CH3)3
MDL Number MFCD00001938
Quantity 10 mg
Molecular Weight (g/mol) 141.17
Percent Purity ≥97.0% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.