CAS RN 111787-83-8
CAS RN 111787-83-8
IUPAC Name:
ethyl 5-(4-fluorophenyl)-2-methylfuran-3-carboxylate
Synonyms:
ethyl 5-(4-fluorophenyl)-2-methylfuran-3-carboxylate
Molecular Weight (g/mol):
248.25
Molecular Formula:
C14H13FO3
InChi Key:
XUYCJNMLONEKAS-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(C)OC(=C1)C1=CC=C(F)C=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Ethyl 5-(4-fluorophenyl)-2-methyl-3-furoate, 95%, Thermo Scientific™
CAS: 111787-83-8 Molecular Formula: C14H13FO3 Molecular Weight (g/mol): 248.25 MDL Number: MFCD05664426 InChI Key: XUYCJNMLONEKAS-UHFFFAOYSA-N Synonym: ethyl 5-4-fluorophenyl-2-methyl-3-furoate,ethyl 5-4-fluorophenyl-2-methylfuran-3-carboxylate,3-furancarboxylic acid,5-4-fluorophenyl-2-methyl-, ethyl ester,acmc-20mesw,2-methyl-5-4-fluorophenyl furan-3-carboxylic acid ethyl ester PubChem CID: 2794727 IUPAC Name: ethyl 5-(4-fluorophenyl)-2-methylfuran-3-carboxylate SMILES: CCOC(=O)C1=C(C)OC(=C1)C1=CC=C(F)C=C1