Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 5-(4-fluorophenyl)-2-methyl-3-furoate, 95%, Thermo Scientific™

Catalog No. MO01270DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO01270DA 1 g Amber Glass Bottle
MO01270CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MO01270DA Supplier Thermo Scientific Chemicals Supplier No. MO01270DA

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 111787-83-8
Molecular Formula C14H13FO3
Molecular Weight (g/mol) 248.25
MDL Number MFCD05664426
InChI Key XUYCJNMLONEKAS-UHFFFAOYSA-N
Synonym ethyl 5-4-fluorophenyl-2-methyl-3-furoate, ethyl 5-4-fluorophenyl-2-methylfuran-3-carboxylate, 3-furancarboxylic acid,5-4-fluorophenyl-2-methyl-, ethyl ester, acmc-20mesw, 2-methyl-5-4-fluorophenyl furan-3-carboxylic acid ethyl ester
PubChem CID 2794727
IUPAC Name ethyl 5-(4-fluorophenyl)-2-methylfuran-3-carboxylate
SMILES CCOC(=O)C1=C(C)OC(=C1)C1=CC=C(F)C=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 248.25
Chemical Name or Material Ethyl 5-(4-fluorophenyl)-2-methyl-3-furoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.