accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COONH2OHO

CAS RN 1119-33-1

IUPAC Name: 2-amino-5-ethoxy-5-oxopentanoic acid
Synonyms: 5-ethyl-L-glutamate
Molecular Weight (g/mol): 175.18
Molecular Formula: C7H13NO4
InChi Key: XMQUEQJCYRFIQS-UHFFFAOYNA-N
SMILES: CCOC(=O)CCC(N)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)