Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamic acid 5-ethyl ester, 95%, Thermo Scientific™

Catalog No. AAH6316806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6316806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6316806 Supplier Thermo Scientific Chemicals Supplier No. H6316806

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
L-Glutamic acid 5-ethyl ester is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in water 100 mg/ml.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119-33-1
Molecular Formula C7H13NO4
Molecular Weight (g/mol) 175.18
MDL Number MFCD00037217
InChI Key XMQUEQJCYRFIQS-UHFFFAOYNA-N
Synonym h-glu oet-oh, l-glutamic acid 5-ethyl ester, l-glutamic acid gamma-ethyl ester, ethyl glutamate, unii-3ec85nwr4k, 5-ethyl-l-glutamate, s-2-amino-5-ethoxy-5-oxopentanoic acid, 2s-2-amino-5-ethoxy-5-oxopentanoic acid, 3ec85nwr4k, l-glutamic acid-5-ethyl ester
PubChem CID 3082305
SMILES CCOC(=O)CCC(N)C(O)=O

Specifications

Melting Point 179°C (decomposition)
Color White
Solubility Information Soluble in water 100mg/ml.
IUPAC Name 2-amino-5-ethoxy-5-oxopentanoic acid
Quantity 5 g
Formula Weight 175.18
Percent Purity 95%
Physical Form Powder
Chemical Name or Material L-Glutamic acid 5-ethyl ester
EINECSNumber 214-274-6
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.