accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1119-62-6

IUPAC Name: 3-[(2-carboxyethyl)disulfanyl]propanoic acid
Synonyms: 4,5-dithia-octandioic acid
Molecular Weight (g/mol): 210.26
Molecular Formula: C6H10O4S2
InChi Key: YCLSOMLVSHPPFV-UHFFFAOYSA-N
SMILES: OC(=O)CCSSCCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)