Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 3,3'-Dithiodipropionic acid

Catalog No. 10901050G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-901-050G 50 g
10-901-0250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-901-050G Supplier Sigma Aldrich Supplier No. 10901050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

3,3'-Dithiodipropionic acid

Specifications

CAS 1119-62-6
Melting Point 155°C to 158°C (lit.)
Molecular Formula C6H10O4S2
Linear Formula HOCOCH2CH2SSCH2CH2COOH
Quantity 50 g
MDL Number MFCD00002780
Synonym 2-Carboxyethyl disulfide; Bis(2-carboxyethyl) disulfide
Molecular Weight (g/mol) 210.27
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.