accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN(Z)

CAS RN 1119-85-3

IUPAC Name: (3Z)-hex-3-enedinitrile
Synonyms: (3Z)-hex-3-enedinitrile
Molecular Weight (g/mol): 106.13
Molecular Formula: C6H6N2
InChi Key: BSVZXPLUMFUWHW-UPHRSURJSA-N
SMILES: N#CC\C=C/CC#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)