Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Hexenedinitrile 98.0+%, TCI America™
SDP

Catalog No. H03575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H03575G 5 g
H035725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H03575G Supplier TCI America Supplier No. H03575G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1119-85-3
Molecular Formula C6H6N2
Molecular Weight (g/mol) 106.13
MDL Number MFCD00001964
InChI Key BSVZXPLUMFUWHW-UPHRSURJSA-N
Synonym 1,4-Dicyano-2-butene
PubChem CID 5324708
IUPAC Name (3Z)-hex-3-enedinitrile
SMILES N#CC\C=C/CC#N

Specifications

Melting Point 76°C
Color White-Yellow
Boiling Point 120°C
Quantity 5 g
UN Number 3439
Formula Weight 106.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Hexenedinitrile
RTECSNumber MP6750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.