accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1119283-74-7

IUPAC Name: tert-butyl N-[4-(4-methylpiperazin-1-yl)cyclohexyl]carbamate
Synonyms: tert-butyl N-[4-(4-methylpiperazin-1-yl)cyclohexyl]carbamate
Molecular Weight (g/mol): 297.44
Molecular Formula: C16H31N3O2
InChi Key: HVPDOBVAGJYPNN-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1CCC(CC1)NC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results