Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1-(Boc-amino)-4-(4-methyl-1-piperazinyl)cyclohexane, 97%, Thermo Scientific™

Catalog No. AAH62731MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH62731MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH62731MD Supplier Thermo Scientific Chemicals Supplier No. H62731MD

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1119283-74-7
Molecular Formula C16H31N3O2
Molecular Weight (g/mol) 297.443
InChI Key HVPDOBVAGJYPNN-UHFFFAOYSA-N
Synonym tert-butyl trans-4-4-methylpiperazin-1-yl cyclohexylcarbamate, tert-butyl n-1r,4r-4-4-methylpiperazin-1-yl cyclohexyl carbamate
PubChem CID 58236010
IUPAC Name tert-butyl N-[4-(4-methylpiperazin-1-yl)cyclohexyl]carbamate
SMILES CC(C)(C)OC(=O)NC1CCC(CC1)N2CCN(CC2)C

Specifications

Quantity 250 mg
Formula Weight 297.43
Percent Purity 97%
Physical Form Solid
Chemical Name or Material trans-1-(Boc-amino)-4-(4-methyl-1-piperazinyl)cyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.