accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3CH3

CAS RN 112-18-5

IUPAC Name: dodecyldimethylamine
Synonyms: dimethyl lauramine
Molecular Weight (g/mol): 213.41
Molecular Formula: C14H31N
InChi Key: YWFWDNVOPHGWMX-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCN(C)C
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)