Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N,N-Dimethyldodecylamine 96.0+%, TCI America™
SDP

Catalog No. D0002500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D0002500ML 500 mL
D000225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D0002500ML Supplier TCI America Supplier No. D0002500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 112-18-5 | Formula: C14H31N | Weight: 213.409 g/mol

Chemical Identifiers

CAS 112-18-5
Molecular Formula C14H31N
Molecular Weight (g/mol) 213.409
MDL Number MFCD00008970
InChI Key YWFWDNVOPHGWMX-UHFFFAOYSA-N
Synonym n,n-dimethyldodecylamine, lauryldimethylamine, dodecyldimethylamine, antioxidant dda, dimethyl lauramine, n,n-dimethyl-n-dodecylamine, 1-dodecanamine, n,n-dimethyl, n,n-dimethyllaurylamine, dda antioxidant, barlene 125
PubChem CID 8168
IUPAC Name N,N-dimethyldodecan-1-amine
SMILES CCCCCCCCCCCCN(C)C

Specifications

Melting Point -20°C
Color Yellow
Boiling Point 149°C
Quantity 500 mL
UN Number 2735
Formula Weight 213.41
Percent Purity ≥96.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material N,N-Dimethyldodecylamine
EINECSNumber (2)-0176
RTECSNumber JR6600000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.