accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNH3

CAS RN 112-20-9

IUPAC Name: nonan-1-aminium chloride
Synonyms: nonan-1-aminium chloride
Molecular Weight (g/mol): 179.73
Molecular Formula: C9H22ClN
InChi Key: IMGHCWMUWVHYCO-UHFFFAOYSA-N
SMILES: [Cl-].CCCCCCCCC[NH3+]
Boiling Point 
  • (3)
  • (2)
  • (5)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (8)
Percent Purity 
  • (3)
  • (2)
  • (2)
  • (6)
Quantity 
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (5)
  • (2)
  • (6)
  • (3)
  • (2)
  • (5)
  • (3)
  • (5)
  • (8)
  • (3)
  • (2)
  • (2)
  • (6)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)