Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Nonylamine

Catalog No. 743985ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-398-5ML 5 mL
74-398-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74-398-5ML Supplier Sigma Aldrich Supplier No. 743985ML
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Nonylamine is an alkylamine generally used to insert c9 chains to substrates
  • It is also used as intercalating material in the exfoliation of titanosilicates and α-layered zirconium phosphate

Specifications

CAS 112-20-9
Density 0.782 g/mL (at 25°C (literature))
Boiling Point 201°C (lit.)
Molecular Formula C9H21N
Linear Formula CH3(CH2)8 NH2
Refractive Index n20/D 1.433 (literature); n20/D 1.433
MDL Number MFCD00008249
Quantity 5 mL
Synonym 1-Aminononane
Molecular Weight (g/mol) 143.27
Percent Purity ≥99.5% (GC)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.