accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH2NH2CH3(E)

CAS RN 112-21-0

IUPAC Name: ethyl[(2E)-4-(ethylazaniumyl)but-2-en-1-yl]azanium
Synonyms: ethyl[(2E)-4-(ethylammonio)but-2-en-1-yl]azanium
Molecular Weight (g/mol): 144.26
Molecular Formula: C8H20N2
InChi Key: YWWSWEIXJXYQJB-AATRIKPKSA-P
SMILES: CC[NH2+]C\C=C\C[NH2+]CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results