accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NHNNHNH2

CAS RN 112-24-3

IUPAC Name: (2-aminoethyl)({2-[(2-aminoethyl)amino]ethyl})amine
Synonyms: trien trientina trientine araldite hardener HY 951 trientine preparation trientine (substance) tecza trientinum araldite HY 951 trientine (product)
Molecular Weight (g/mol): 146.24
Molecular Formula: C6H18N4
InChi Key: VILCJCGEZXAXTO-UHFFFAOYSA-N
SMILES: NCCNCCNCCN
Molecular Weight (g/mol) 
  • (6)
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (1)
  • (2)
  • (9)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (2)
  • (6)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (6)
  • (2)
  • (6)
  • (6)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (9)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (6)
  • (3)
  • (6)
  • (5)