Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylenetetramine, Thermo Scientific™

Catalog No. AC157925000
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC157925000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC157925000 Supplier Thermo Scientific Chemicals Supplier No. 157925000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-24-3
Molecular Formula C6H18N4
Molecular Weight (g/mol) 146.238
InChI Key VILCJCGEZXAXTO-UHFFFAOYSA-N
Synonym triethylenetetramine, trientine, trien, teta, tecza, triethylene tetramine, araldite hardener hy 951, 1,2-ethanediamine, n,n'-bis 2-aminoethyl, deh 24, araldite hy 951
PubChem CID 5565
ChEBI CHEBI:39501
IUPAC Name N'-[2-(2-aminoethylamino)ethyl]ethane-1,2-diamine
SMILES C(CNCCNCCN)N

Specifications

Density 0.98g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Refractive Index 1.4961 to 1.4991 (20°C, 589nm)
Linear Formula H2NCH2CH2NHCH2CH2NHCH2CH2NH2
Quantity 500 g
Beilstein 04, 255
Fieser 01,1204
Merck Index 15, 9828
Formula Weight 146.24
Percent Purity ≥94%
Physical Form Liquid
Chemical Name or Material Triethylenetetramine, 95%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS P Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Harmful in contact with skin.
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.