accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOH

CAS RN 112-25-4

IUPAC Name: 2-(hexyloxy)ethan-1-ol
Synonyms: hexyloxyethanol
Molecular Weight (g/mol): 146.23
Molecular Formula: C8H18O2
InChi Key: UPGSWASWQBLSKZ-UHFFFAOYSA-N
SMILES: CCCCCCOCCO
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)