accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClOOCl

CAS RN 112-26-5

IUPAC Name: 1-chloro-2-[2-(2-chloroethoxy)ethoxy]ethane
Synonyms: 1,2-bis(2-chloroethoxy)ethane
Molecular Weight (g/mol): 187.06
Molecular Formula: C6H12Cl2O2
InChi Key: AGYUOJIYYGGHKV-UHFFFAOYSA-N
SMILES: ClCCOCCOCCCl
Boiling Point 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)