accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112-36-7

IUPAC Name: 1-ethoxy-2-(2-ethoxyethoxy)ethane
Synonyms: bis (2-ethoxy ethyl) ether
Molecular Weight (g/mol): 162.23
Molecular Formula: C8H18O3
InChi Key: RRQYJINTUHWNHW-UHFFFAOYSA-N
SMILES: CCOCCOCCOCC
Molecular Weight (g/mol) 
  • (7)
  • (3)
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (8)
  • (7)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (6)
  • (4)
  • (1)