CAS RN 112-48-1
IUPAC Name:
1-(2-butoxyethoxy)butane
Synonyms:
1,2-dibutoxyethane acetaldehyde, dibutyl acetal
Molecular Weight (g/mol):
174.28
Molecular Formula:
C10H22O2
InChi Key:
GDXHBFHOEYVPED-UHFFFAOYSA-N
SMILES:
CCCCOCCOCCCC
1
–
1
of
1
results
1
Ethylene Glycol Dibutyl Ether 98.0+%, TCI America™
CAS: 112-48-1 Molecular Formula: C10H22O2 Molecular Weight (g/mol): 174.28 MDL Number: MFCD00048785 InChI Key: GDXHBFHOEYVPED-UHFFFAOYSA-N Synonym: 1,2-Dibutoxyethane, Dibutyl Cellosolve, Dibutyl Glycol PubChem CID: 8188 IUPAC Name: 1-(2-butoxyethoxy)butane SMILES: CCCCOCCOCCCC