accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112-51-6

IUPAC Name: 1-(pentyldisulfanyl)pentane
Synonyms: 6,7-dithiadodecane
Molecular Weight (g/mol): 206.41
Molecular Formula: C10H22S2
InChi Key: YSQZSPCQDXHJDJ-UHFFFAOYSA-N
SMILES: CCCCCSSCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results