Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diamyl Disulfide 95.0+%, TCI America™
SDP

Catalog No. D013025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D013025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D013025ML Supplier TCI America Supplier No. D013025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 112-51-6
Molecular Formula C10H22S2
Molecular Weight (g/mol) 206.41
MDL Number MFCD00039964
InChI Key YSQZSPCQDXHJDJ-UHFFFAOYSA-N
Synonym Amyl Disulfide, Dipentyl Disulfide, Pentyl Disulfide
PubChem CID 8191
IUPAC Name 1-(pentyldisulfanyl)pentane
SMILES CCCCCSSCCCCC

Specifications

Boiling Point 142°C
Quantity 25 mL
Formula Weight 206.41
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Diamyl Disulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.