accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112-55-0

IUPAC Name: dodecane-1-thiol
Synonyms: dodecyl mercaptan
Molecular Weight (g/mol): 202.4
Molecular Formula: C12H26S
InChi Key: WNAHIZMDSQCWRP-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCS
Boiling Point 
  • (4)
  • (3)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (7)
Percent Purity 
  • (2)
  • (4)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (4)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (2)
  • (4)
  • (7)
  • (2)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (7)