Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Dodecanethiol

Catalog No. 471364500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 L
18 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-136-4500ML 500 mL
47-136-42L 2 L
47-136-418L 18 L
47-136-4100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 47-136-4500ML Supplier Sigma Aldrich Supplier No. 471364500ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Dodecanethiol (DDT) is an alkyl thiol that forms a self-assembled monolayer (SAM) and can be used as an organic source of sulfur with balanced physio-chemical properties.This molecule is used for the production of hydrophobic SAMs. It can also be used in mixed SAMs to give a hydrophobic background and act as a spacer to move other functional groups or domains farther apart.Product of Arkema Inc.

Specifications

CAS 112-55-0
Density 0.845 g/mL (at 25°C)
Boiling Point 266°C to 283°C
Molecular Formula C12H26S
Refractive Index n20/D 1.459 (literature)
Linear Formula CH3(CH2)11 SH
Quantity 500 mL
MDL Number MFCD00004885
Synonym n-Dodecyl mercaptan; Dodecyl mercaptan; Lauryl mercaptan; Mercaptan C12; NDM
Molecular Weight (g/mol) 202.4
Percent Purity ≥98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.