accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3CH3

CAS RN 112-75-4

IUPAC Name: dimethyl(tetradecyl)amine
Synonyms: dimethyl myristamine
Molecular Weight (g/mol): 241.46
Molecular Formula: C16H35N
InChi Key: SFBHPFQSSDCYSL-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCN(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)