accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH

CAS RN 112-92-5

IUPAC Name: octadecan-1-ol
Synonyms: stearyl alcohol 1-0ctadecanol lorol 28 alcohol stearylicus steraffine octadecanol NF stearyl alcohol pure aldol 62 siponol SC polaax
Molecular Weight (g/mol): 270.5
Molecular Formula: C18H38O
InChi Key: GLDOVTGHNKAZLK-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCO
Boiling Point 
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (4)
  • (1)
  • (2)
  • (5)
  • (2)
  • (2)
  • (1)
  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (4)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)