Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stearyl Alcohol, NF, 90-102%, Spectrum™ Chemical
SDP

Catalog No. 18607017 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-017 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-017 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ST115500GM

missing translation for 'validMainframeMsg'

ST115, 112-92-5, C18H38O

Spectrum™ Chemical Stearyl Alcohol, NF is used as an emulsion stabilizer, fragrance ingredient, emulsifying agent, foam booster, and as a viscosity increasing agent. All Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 112-92-5
Molecular Formula C18H38O
Molecular Weight (g/mol) 270.50
InChI Key GLDOVTGHNKAZLK-UHFFFAOYSA-N
IUPAC Name octadecan-1-ol
SMILES CCCCCCCCCCCCCCCCCCO

Specifications

CAS Max % 1
Boiling Point 210°C (410°F)
Ignition Residue 0.001
Packaging Poly Bottle
Quantity 500 g
Percent Purity 90 to 102%
Grade NF
Physical Form Powder or Waxy Solid
Chemical Name or Material Stearyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.