accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1120-02-1

IUPAC Name: trimethyl(octadecyl)azanium bromide
Synonyms: steartrimonium bromide
Molecular Weight (g/mol): 392.51
Molecular Formula: C21H46BrN
InChi Key: SZEMGTQCPRNXEG-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCCCCCCCCCCCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results