Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trimethylstearylammonium Bromide 98.0+%, TCI America™
SDP

Catalog No. S036425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S036425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S036425G Supplier TCI America Supplier No. S036425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1120-02-1
Molecular Formula C21H46BrN
Molecular Weight (g/mol) 392.51
MDL Number MFCD00043171
InChI Key SZEMGTQCPRNXEG-UHFFFAOYSA-M
Synonym octadecyltrimethylammonium bromide, n,n,n-trimethyloctadecan-1-aminium bromide, steartrimonium bromide, stearyltrimethylammonium bromide, trimethyloctadecylammonium bromide, octmab, unii-bpw1dd4iz8, octadecy trimethyl ammonium bromide, 1-octadecanaminium, n,n,n-trimethyl-, bromide, bpw1dd4iz8
PubChem CID 70708
IUPAC Name trimethyl(octadecyl)azanium;bromide
SMILES CCCCCCCCCCCCCCCCCC[N+](C)(C)C.[Br-]

Specifications

Color White
Quantity 25 g
Formula Weight 392.51
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Trimethylstearylammonium Bromide
EINECSNumber (2)-0184&(9)-1971
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.