accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1120-06-5

IUPAC Name: decan-2-ol
Synonyms: decan-2-ol
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: ACUZDYFTRHEKOS-UHFFFAOYNA-N
SMILES: CCCCCCCCC(C)O
Molecular Weight (g/mol) 
  • (2)
  • (5)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (2)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (6)
  • (2)
  • (4)
  • (4)