Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Decanol, MP Biomedicals

Catalog No. ICN210853 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210853 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210853 Supplier MP Biomedicals, Inc Supplier No. 0521085305
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1120-06-5
Molecular Formula C10H22O
Molecular Weight (g/mol) 158.285
InChI Key ACUZDYFTRHEKOS-UHFFFAOYSA-N
Synonym 2-decanol, 2-hydroxydecane, 2-decyl alcohol, methyl-n-octylcarbinol, decanol-2, methyl-n-octyl carbinol, acmc-1bovc, chembl46477
PubChem CID 14254
IUPAC Name decan-2-ol
SMILES CCCCCCCCC(C)O

Specifications

Quantity 5 g
Formula Weight 158.29
Chemical Name or Material 2-Decanol
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.